Dimethyl fumarate, 99%
Dimethyl fumarate, 99%
Dimethyl fumarate, 99%
Thermo Scientific Chemicals

Dimethyl fumarate, 99%

CAS: 624-49-7 | C6H8O4 | 144.126 g/mol
Quantity:
25 g
100 g
250 g
Catalog number A10402.14
also known as A10402-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS624-49-7
IUPAC Name1,4-dimethyl (2E)-but-2-enedioate
Molecular FormulaC6H8O4
InChI KeyLDCRTTXIJACKKU-ONEGZZNKSA-N
SMILESCOC(=O)\C=C\C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or crystalline powder or flakes
Free acid (titration)≤1.5% as fumaric acid
Appearance (Color)White to cream to pale brown
Identification (FTIR)Conforms
Assay (GC)≥98.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Dimethyl fumarate acts as an immunomodulator. It is also used in cycloaddition reactions involving ylides, benzenes and amino acids. It is added to food grains,tobacco,leather and preservation. Clothing

Solubility
Soluble in water (1.6 mg/ml at 20°C), methanol (30-36 mg/ml), ethanol (10 mg/ml at 25°C), DMSO (29 mg/ml at 25°C), and DMF (∼12 mg/ml).

Notes
Do not store together with acids. Incompatible with acids, bases, oxidizing agents, reducing agents. Store at room temperature.
RUO – Research Use Only
BW van Oosten; J Killestein; F Barkhof; Chris H. Polman,; Mike P. Wattjes. PML in a Patient Treated with Dimethyl Fumarate from a Compounding Pharmacy. N Engl J Med. 2013, 368 1658-1659.