Caffeine, 99%
Caffeine, 99%
Caffeine, 99%
Thermo Scientific Chemicals

Caffeine, 99%

CAS: 58-08-2 | C8H10N4O2 | 194.19 g/mol
Quantity:
100 g
250 g
1000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A10431.30
also known as A10431-30
Price (JPY)
-
Request A Quote
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS58-08-2
IUPAC Name1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular FormulaC8H10N4O2
InChI KeyRYYVLZVUVIJVGH-UHFFFAOYSA-N
SMILESCN1C=NC2=C1C(=O)N(C)C(=O)N2C
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Appearance (Color)White to pale cream
Identification (FTIR)Conforms
This material is traded for research purposes ONLY. Thermo Fisher does NOT certify this material as suitable for pharmaceutical, cosmetic, veterinary or food use. Caffeine has inhibitory action against lung and colon tumorigenses and virus induction.Caffeine is used to treat and prevent bronchopulmonary dysplasia in premature infants. It is also used for the treatment of apnea of prematurity and orthostatic hypotension. It is widely used as an additive in food, cosmetics, pharmaceuticals and beverages. It confers a protective effect against some diseases including Parkinson's disease and certain types of cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in pyrrole, tetrahydrofuran+ water mixture and ethyl acetate. Slightly soluble in water and ethanol.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Elias V. E. F.; Coso, J. D.; Hamouti, N.; Ortega, JF.; Munoz, G.; Guerra J. M.; Rodriguez R. M. Ingestion of a moderately high caffeine dose before exercise increases postexercise energy expenditure. Int. J. Sport Nutr. Exerc. Metab. 2015, 25 (1), 46-53.
  2. Nakayama, F.; Mizuno, K.; Kato, M. Biosynthesis of caffeine underlying the diversity of motif B' methyltransferase. Nat Prod Commun 10 (5), 799-801.