Dibromomethane, 99%
Dibromomethane, 99%
Dibromomethane, 99%
Thermo Scientific Chemicals

Dibromomethane, 99%

CAS: 74-95-3 | CH2Br2 | 173.835 g/mol
Quantity:
100 g
500 g
2500 g
Catalog number A10456.22
also known as A10456-22
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS74-95-3
IUPAC Namedibromomethane
Molecular FormulaCH2Br2
InChI KeyFJBFPHVGVWTDIP-UHFFFAOYSA-N
SMILESBrCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.5%
Refractive Index1.5395-1.5425 @ 20?C
FormLiquid
Dibromomethane is used as solvent in organic synthesis. It acts as an intermediate in the manufacture of specialty chemicals, agrochemicals and pharmaceuticals. It is useful as extractant and utilized for the determination of 5-nitroimidazoles (5-NDZ) in environmental waters. It is involved in the convertion of catechols to their methylenedioxy derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibromomethane is used as solvent in organic synthesis. It acts as an intermediate in the manufacture of specialty chemicals, agrochemicals and pharmaceuticals. It is useful as extractant and utilized for the determination of 5-nitroimidazoles (5-NDZ) in environmental waters. It is involved in the convertion of catechols to their methylenedioxy derivatives.

Solubility
Miscible with water, chloroform, acetone, ether and alcohol.

Notes
Store in cool place. Incompatible with strong oxidizing agents, aluminum and magnesium.
RUO – Research Use Only

General References:

  1. For in situ formation of dibromomethyllithium by reaction with lithiated 2,2,6,6-Tetramethyl piperidine, A18712, and use in a one-pot ester homologation sequence, safer and more convenient than the classical Arndt-Eistert (diazomethane) method, see: Org. Synth. Coll., 9, 426 (1998):
  2. Reaction with NaHMDS generates monobromocarbene, which reacts with alkenes to give bromocyclopropanes: Synthesis, 201 (1972). It can also be generated by reaction with diethylzinc in the presence of O2: J. Chem. Soc., Chem. Commun., 364 (1975).
  3. The Simmons-Smith reaction (see Diiodomethane, A15457) may be extended to the use of dibromomethane in the cyclopropanation of simple electron-deficient alkenes, using a reducing agent prepared from NiBr2, NaI and Zn: Bull. Chem. Soc. Jpn., 56, 1025 (1983); or with CuCl/Zn: J. Org. Chem., 55, 2491 (1990).
  4. With TiCl4/Zn, methylenation of ketones occurs, as a useful mild alternative to the Wittig reaction: Tetrahedron Lett., 23, 4293 (1982); Org. Synth. Coll., 8, 386 (1993). For a variant using Zn, CuCl and catalytic TiCl4, see: J. Org. Chem., 54, 2388 (1989).
  5. With alcohols, formaldehyde acetals can be formed under phase-transfer conditions: Bull. Chem. Soc. Jpn., 66, 2149 (1993). Diols give methylenedioxy derivatives, which are components of many biologically-important molecules. For use of KF in DMF as base in the methylenation of catechols, see: Tetrahedron Lett., 3361 (1976). KOH in DMSO has been recommended for the methylenation of base-stable carbohydrate diols: Synthesis, 421 (1982). For the methylenation of ribonucleosides under phase-transfer conditions, see: Synthesis, 715 (1981).