ACK Lysing Buffer - FAQs
ACK Lysing Buffer - FAQs
View additional product information for ACK Lysing Buffer - FAQs (A1049201)
3 product FAQs found
What alternative do you offer for ACK Lysing Buffer (Cat. No. A1049201)?
Alternative products we offer are eBioscience 1X RBC Lysis Buffer (Cat. No. 00-4333-57) or eBioscience 10X RBC Lysis Buffer (Multi-species) (Cat. No. 00-4300-54).
Can I use ACK Lysing Buffer with heparinized blood?
Yes, ACK Lysing Buffer can be used with heparinized blood. The principle of lysis is osmotic treatment.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How can I use ACK Lysing Buffer to lyse red blood cells?
A volume of ACK lysing buffer corresponding to 10-20 times the volume of the blood sample is sufficient to lyse red blood cells.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.