3,4-Dibromothiophene, 98+%
3,4-Dibromothiophene, 98+%
3,4-Dibromothiophene, 98+%
Thermo Scientific Chemicals

3,4-Dibromothiophene, 98+%

CAS: 3141-26-2 | C4H2Br2S | 241.93 g/mol
Quantity:
5 g
25 g
Catalog number A10512.14
also known as A10512-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS3141-26-2
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Refractive Index1.6375-1.6415 @ 20°C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only