2,4-Dichloroaniline, 99%
2,4-Dichloroaniline, 99%
2,4-Dichloroaniline, 99%
2,4-Dichloroaniline, 99%
Thermo Scientific Chemicals

2,4-Dichloroaniline, 99%

CAS: 554-00-7 | C6H5Cl2N | 162.013 g/mol
Quantity:
100 g
250 g
500 g
2500 g
Catalog number A10518.0E
also known as A10518-0E
Price (JPY)
-
Request A Quote
Quantity:
2500 g
Request bulk or custom format
Chemical Identifiers
CAS554-00-7
IUPAC Name2,4-dichloroaniline
Molecular FormulaC6H5Cl2N
InChI KeyKQCMTOWTPBNWDB-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
CommentMay darken on storage
Appearance (Color)White to cream to grey or pale brown to brown
FormPowder or crystalline powder or crystals or flakes and/or chunks
Melting Point (clear melt)58-67°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only