Methyl salicylate, 98%
Methyl salicylate, 98%
Methyl salicylate, 98%
Thermo Scientific Chemicals

Methyl salicylate, 98%

CAS: 119-36-8 | C8H8O3 | 152.149 g/mol
Quantity:
250 g
500 g
2500 g
Catalog number A10578.0E
also known as A10578-0E
Price (JPY)
-
Quantity:
2500 g
Request bulk or custom format
Chemical Identifiers
CAS119-36-8
IUPAC Namemethyl 2-hydroxybenzoate
Molecular FormulaC8H8O3
InChI KeyOSWPMRLSEDHDFF-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5335-1.5375 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
Assay (GC)≥97.5%
FormLiquid
Methyl salicylate is used as a flavoring agent and providing fragrance to products like chewing gum and mints. It acts as an anticoagulant by interruption of the vitamin K1-epoxide cycle. It is also used in high concentrations as a rubefacient to treat joint, muscular pain and acute pain. It is utilized as an anti-herbivore defense system in various plants. It is used as a transfer agent to get a manual copy of an image on a surface and to clear plant or animal tissue samples to be used for microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl salicylate is used as a flavoring agent and providing fragrance to products like chewing gum and mints. It acts as an anticoagulant by interruption of the vitamin K1-epoxide cycle. It is also used in high concentrations as a rubefacient to treat joint, muscular pain and acute pain. It is utilized as an anti-herbivore defense system in various plants. It is used as a transfer agent to get a manual copy of an image on a surface and to clear plant or animal tissue samples to be used for microscopy.

Solubility
Miscible with water, chloroform, ethanol, methonol and ether.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Valverde, J. M.; Giménez, M. J.; Guillén, F.; Valero, D.; Martínez-Romero, D.; Serrano, M. Methyl salicylate treatments of sweet cherry trees increase antioxidant systems in fruit at harvest and during storage. Postharvest Biol. Technol. 2015, 109, 106-113.
  2. Zhou, P.; Hoffmann, M. R.; Han, K.; He, G. New Insights into the Dual Fluorescence of Methyl Salicylate: Effects of Intermolecular Hydrogen Bonding and Solvation. J. Phys. Chem. B 2014, 119 (6), 2125-2131.