Thermo Scientific Chemicals

DL-Threonine, 99%

Catalog number: A10606.36
500 g, Each
Thermo Scientific Chemicals

DL-Threonine, 99%

Catalog number: A10606.36
500 g, Each
Quantity
Catalog number: A10606.36
also known as A10606-36
Price (USD)
Quantity
-

Chemical Identifiers

CAS
80-68-2
IUPAC Name
(2R,3R)-2-amino-3-hydroxybutanoic acid
Molecular Formula
C4H9NO3
InChI Key
AYFVYJQAPQTCCC-PWNYCUMCSA-N
SMILES
C[C@@H](O)[C@@H](N)C(O)=O
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to pale cream
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

DL-Threonine is used as a polar essential amino acid. It serves as a precursor of glycine as well as used in the biosynthesis of proteins. Further, it is used for biochemical research. In addition to this, it is used as a nutrition enhancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Threonine is used as a polar essential amino acid. It serves as a precursor of glycine as well as used in the biosynthesis of proteins. Further, it is used for biochemical research. In addition to this, it is used as a nutrition enhancer.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text