Thermo Scientific Chemicals

3-Iodotoluene, 99%, Thermo Scientific Chemicals

50 g, Each
Thermo Scientific Chemicals

3-Iodotoluene, 99%, Thermo Scientific Chemicals

50 g, Each
Quantity
Catalog number: A10999.18
also known as A10999-18
Price (USD)
Price: 74.70
Your Price:
Quantity
-

Chemical Identifiers

CAS
625-95-6
IUPAC Name
1-iodo-3-methylbenzene
Molecular Formula
C7H7I
InChI Key
VLCPISYURGTGLP-UHFFFAOYSA-N
SMILES
CC1=CC=CC(I)=C1
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Form
Liquid
Refractive Index
1.6035-1.6065 @ 20?C

Description

3-Iodotoluene was used in the preparartion of indene derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodotoluene was used in the preparartion of indene derivatives.

Solubility
Not miscible in water. Soluble in benzene, alcohol and ether.

Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only