2-Ethyl-3-methylpyrazine, 98+%
2-Ethyl-3-methylpyrazine, 98+%
2-Ethyl-3-methylpyrazine, 98+%
Thermo Scientific Chemicals

2-Ethyl-3-methylpyrazine, 98+%

CAS: 15707-23-0 | C7H10N2 | 122.17 g/mol
Quantity:
5 g
25 g
100 g
Catalog number A11092.22
also known as A11092-22
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS15707-23-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.0%
Refractive Index1.5000-1.5040 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only