2,6-Dichlorobenzaldoxime, 97%
2,6-Dichlorobenzaldoxime, 97%
2,6-Dichlorobenzaldoxime, 97%
Thermo Scientific Chemicals

2,6-Dichlorobenzaldoxime, 97%

CAS: 25185-95-9 | C7H5Cl2NO | 190.02 g/mol
Quantity:
5 g
25 g
100 g
Catalog number A11351.14
also known as A11351-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS25185-95-9
IUPAC Name(E)-N-[(2,6-dichlorophenyl)methylidene]hydroxylamine
Molecular FormulaC7H5Cl2NO
InChI KeyYBSXDWIAUZOFFV-ONNFQVAWSA-N
SMILESO\N=C\C1=C(Cl)C=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)144.0-154.0?C
Appearance (Color)White
Assay (HPLC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only