4-Chloro-3-nitrotoluene, 97+%
4-Chloro-3-nitrotoluene, 97+%
4-Chloro-3-nitrotoluene, 97+%
4-Chloro-3-nitrotoluene, 97+%
Thermo Scientific Chemicals

4-Chloro-3-nitrotoluene, 97+%

CAS: 89-60-1 | C7H6ClNO2 | 171.58 g/mol
Quantity:
100 g
500 g
Catalog number A11383.22
also known as A11383-22
Price (JPY)
-
Request A Quote
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS89-60-1
IUPAC Name1-chloro-4-methyl-2-nitrobenzene
Molecular FormulaC7H6ClNO2
InChI KeyNWESJZZPAJGHRZ-UHFFFAOYSA-N
SMILESCC1=CC=C(Cl)C(=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to orange
Assay (GC)≥97.0%
FormLiquid
Refractive Index1.5550-1.5600 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only