4-Bromophenyl methyl sulfone, 98+%
4-Bromophenyl methyl sulfone, 98+%
4-Bromophenyl methyl sulfone, 98+%
Thermo Scientific Chemicals

4-Bromophenyl methyl sulfone, 98+%

CAS: 3466-32-8 | C7H7BrO2S | 235.095 g/mol
Quantity:
10 g
50 g
Catalog number A11427.09
also known as A11427-09
Price (JPY)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS3466-32-8
IUPAC Name1-bromo-4-methanesulfonylbenzene
Molecular FormulaC7H7BrO2S
InChI KeyFJLFSYRGFJDJMQ-UHFFFAOYSA-N
SMILESCS(=O)(=O)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)99.0-105.0?C
Appearance (Color)White
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only