4-Chloro-3-nitrobenzotrifluoride, 98%
4-Chloro-3-nitrobenzotrifluoride, 98%
4-Chloro-3-nitrobenzotrifluoride, 98%
4-Chloro-3-nitrobenzotrifluoride, 98%
Thermo Scientific Chemicals

4-Chloro-3-nitrobenzotrifluoride, 98%

CAS: 121-17-5 | C7H3ClF3NO2 | 225.55 g/mol
Quantity:
100 g
500 g
2500 g
Catalog number A11488.0E
also known as A11488-0E
Price (JPY)
-
Request A Quote
Quantity:
2500 g
Request bulk or custom format
Chemical Identifiers
CAS121-17-5
IUPAC Name1-chloro-2-nitro-4-(trifluoromethyl)benzene
Molecular FormulaC7H3ClF3NO2
InChI KeyTZGFQIXRVUHDLE-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(=CC=C1Cl)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Refractive Index1.4880-1.4920 @ 20?C
Assay (GC)≥97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only