Benzyl carbamate, 99%
Benzyl carbamate, 99%
Benzyl carbamate, 99%
Thermo Scientific™

Benzyl carbamate, 99%

CAS: 621-84-1 | C8H9NO2 | 151.17 g/mol
Quantity:
25 g
100 g
250 g
Catalog number A11569.22
also known as A11569-22
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS621-84-1
IUPAC Namebenzyl carbamate
Molecular FormulaC8H9NO2
InChI KeyPUJDIJCNWFYVJX-UHFFFAOYSA-N
SMILESNC(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤0.75%
Assay (GC)≥98.5%
Melting Point (clear melt)83-91?C
Appearance (Color)White to pale cream
FormCrystalline powder or flakes
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only