1-(Phenylsulfonyl)pyrrole, 98%
1-(Phenylsulfonyl)pyrrole, 98%
1-(Phenylsulfonyl)pyrrole, 98%
Thermo Scientific Chemicals

1-(Phenylsulfonyl)pyrrole, 98%

CAS: 16851-82-4 | C10H9NO2S | 207.25 g/mol
Quantity:
5 g
25 g
Catalog number A11599.14
also known as A11599-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS16851-82-4
IUPAC Name1-(benzenesulfonyl)-1H-pyrrole
Molecular FormulaC10H9NO2S
InChI KeyPPPXRIUHKCOOMU-UHFFFAOYSA-N
SMILESO=S(=O)(N1C=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormCrystalline powder
Appearance (Color)White to pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only