Thermo Scientific Chemicals

Diethylamine, 99+%, Thermo Scientific Chemicals

Catalog number: A11716.0F
2500 mL, Each
Thermo Scientific Chemicals

Diethylamine, 99+%, Thermo Scientific Chemicals

Catalog number: A11716.0F
2500 mL, Each
Quantity

Chemical Identifiers

CAS
109-89-7
IUPAC Name
diethylamine
Molecular Formula
C4H11N
InChI Key
HPNMFZURTQLUMO-UHFFFAOYSA-N
SMILES
CCNCC
Form
Liquid
Refractive Index
1.3835-1.3865 @ 20?C or 1.3810-1.3850 @ 25?C (UK-sourced material)
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms (UK-sourced material)
Comment
Material sourced in UK and US

Description

Diethylamine is used as a corrosion inhibitor. It is also used in the production of rubber, resins, dyes and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylamine is used as a corrosion inhibitor. It is also used in the production of rubber, resins, dyes and pharmaceuticals. It is also used to cook lysergic acid diethylamide (LSD) and is strictly watched by Drug Enforcement Administration (DEA).

Solubility
Miscible with water, alcohol, ether, chloroform and carbon terachloride.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text