2,4,6-Trimethylbenzoic acid, 99%
2,4,6-Trimethylbenzoic acid, 99%
2,4,6-Trimethylbenzoic acid, 99%
Thermo Scientific Chemicals

2,4,6-Trimethylbenzoic acid, 99%

CAS: 480-63-7 | C10H12O2 | 164.20 g/mol
Quantity:
10 g
50 g
250 g
Catalog number A11831.18
also known as A11831-18
Price (JPY)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS480-63-7
IUPAC Name2,4,6-trimethylbenzoic acid
Molecular FormulaC10H12O2
InChI KeyFFFIRKXTFQCCKJ-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(C(O)=O)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Assay (Silylated GC)≥98.5%
Identification (FTIR)Conforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only