Thiophene-2-carbonitrile, 98%
Thiophene-2-carbonitrile, 98%
Thiophene-2-carbonitrile, 98%
Thermo Scientific Chemicals

Thiophene-2-carbonitrile, 98%

CAS: 1003-31-2 | C5H3NS | 109.146 g/mol
Quantity:
25 g
100 g
500 g
Catalog number A11930.36
also known as A11930-36
Price (JPY)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS1003-31-2
IUPAC Namethiophene-2-carbonitrile
Molecular FormulaC5H3NS
InChI KeyCUPOOAWTRIURFT-UHFFFAOYSA-N
SMILESN#CC1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5620-1.5670 @ 20°C
Thiophene-2-carbonitrile is used as an intermediate in synthetic chemistry and in the pharmaceutical industry. It is also used in the preparation of thiaplatinacycles and 2,2'-thienylpyrroles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene-2-carbonitrile is used as an intermediate in synthetic chemistry and in the pharmaceutical industry. It is also used in the preparation of thiaplatinacycles and 2,2′-thienylpyrroles.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Jiang, T. S.; Wang, G. W. Synthesis of 2-Acylthiophenes by Palladium-Catalyzed Addition of Thiophenes to Nitriles. Adv. Synth. Catal. 2014, 356 (2-3), 369-373.
  2. Rodrigues, A. D.; Imberdis, T.; Perrier, V.; Robitzer, M. Improved synthesis of a quaterthiophene-triazine-diamine derivative, a promising molecule to study pathogenic prion proteins. Tetrahedron Lett. 2015, 56 (2), 368-373.