Thermo Scientific Chemicals

Calcium dihydrogen phosphate hydrate, 97%, Thermo Scientific Chemicals

Catalog number: A12123.36
500 g, Each
Thermo Scientific Chemicals

Calcium dihydrogen phosphate hydrate, 97%, Thermo Scientific Chemicals

Catalog number: A12123.36
500 g, Each
Quantity
Catalog number: A12123.36
also known as A12123-36
Price (USD)
Quantity
-

Chemical Identifiers

CAS
301524-28-7
IUPAC Name
calcium didihydrogen phosphate
Molecular Formula
CaH4O8P2
InChI Key
YYRMJZQKEFZXMX-UHFFFAOYSA-L
SMILES
[Ca++].OP(O)([O-])=O.OP(O)([O-])=O
Form
Powder
Elemental analysis by ICP
Calcium (Ca): 16.46-17.79%
Appearance (Color)
White
Assay (unspecified)
(Based on Ca content): >96.0 to <104.0% (dry wt. basis)
Loss on Drying
≤10% (105?C/vacuum/4hrs)

Description

Calcium dihydrogen phosphate hydrate is a pH control reagent, stabilizer for plastics, manufacture of glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium dihydrogen phosphate hydrate is a pH control reagent, stabilizer for plastics, manufacture of glass.

Solubility
Soluble in water 18 g/L @ 30°C.

Notes
Loses H{2}O at 100°, dec. at 200°
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text