3,5-Dimethylisoxazole, 99%
3,5-Dimethylisoxazole, 99%
3,5-Dimethylisoxazole, 99%
Thermo Scientific Chemicals

3,5-Dimethylisoxazole, 99%

CAS: 300-87-8 | C5H7NO | 97.117 g/mol
Quantity:
25 g
100 g
Catalog number A12217.14
also known as A12217-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS300-87-8
IUPAC Name3,5-dimethyl-1,2-oxazole
Molecular FormulaC5H7NO
InChI KeyFICAQKBMCKEFDI-UHFFFAOYSA-N
SMILESCC1=CC(C)=NO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.5%
Refractive Index1.4400-1.4430 @ 20?C
FormLiquid
The optimization of 3,5-dimethylisoxazole derivatives to develop potent inhibitors of the BET (bromodomain and extra terminal domain) bromodomain family with good ligand efficiency. It has hypoglycemic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The optimization of 3,5-dimethylisoxazole derivatives to develop potent inhibitors of the BET (bromodomain and extra terminal domain) bromodomain family with good ligand efficiency. It has hypoglycemic activity.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Hewings DS; Fedorov O; Filippakopoulos P.; Martin S.; Picaud S.; Tumber A.; Wells C.; Olcina MM.; Freeman K.; Gill A.; Ritchie AJ.; Sheppard DW.; Russell AJ.; Hammond EM; Knapp S; Brennan PE; Conway SJ.Optimization of 3,5-dimethylisoxazole derivatives as potent bromodomain ligands. J Med Chem. 2013, 56 (8), 3217-27.
  2. William E. Dulin; George C. Gerritsen. Hypoglycemic Activity of 3,5 Dimethylisoxazole. Exp Biol Med. 1963, 113 (3), 683-685.
  3. Lithiation occurs first at the 5-methyl group. Subsequent reaction with electrophiles can be followed by further lithiation of the 3-methyl, providing a route to substituted isoxazoles, and also to 1,3-diketones by acid hydrolysis or ß-aminoketones by hydrogenolysis: Tetrahedron Lett., 22, 3699 (1981):
  4. Review: Reaction of 3,5-dimethylisoxazole with some electrophiles: Heterocycles, 6, 805 (1977).