L-Serine methyl ester hydrochloride, 98%
L-Serine methyl ester hydrochloride, 98%
L-Serine methyl ester hydrochloride, 98%
Thermo Scientific Chemicals

L-Serine methyl ester hydrochloride, 98%

CAS: 5680-80-8 | C4H10ClNO3 | 155.578 g/mol
Quantity:
5 g
25 g
100 g
Catalog number A12355.14
also known as A12355-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS5680-80-8
IUPAC Namemethyl 2-amino-3-hydroxypropanoate hydrochloridyl
Molecular FormulaC4H9ClNO3
InChI KeyQDXGLJWDLGMECL-UHFFFAOYNA-N
SMILES[Cl].COC(=O)C(N)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)> 97.5%
Water Content (Karl Fischer Titration)< 1.0%
Optical Rotation+5 ?0.5? (c=2 in methanol)
Appearance (Color)White
Formcrystalline powder
L-Serine methyl ester hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields. It is also a used as a raw material in peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Serine methyl ester hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields. It is also a used as a raw material in peptide synthesis.

Solubility
Soluble in water (50 mg/ml), and methanol

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
RUO – Research Use Only

General References:

  1. Mukund P. Sibi.; Prasad K. Deshpande.; Anthony J. La Loggia.; James W. Christensen. Synthesis of N-BOC-D-diphenylalanine from L-serine methyl ester hydrochloride. Tetrahedron Letters. 1995, 36 (49), 8961-8964.
  2. James S. Nowick.; Noel A. Powell.; Tram M. Nguyen.; Glenn Noronha. An improved method for the synthesis of enantiomerically pure amino acid ester isocyanates. J. Org. Chem. 1992, 57 (26), 7364-7366.