5-Nitrouracil, 98+%
5-Nitrouracil, 98+%
5-Nitrouracil, 98+%
Thermo Scientific Chemicals

5-Nitrouracil, 98+%

CAS: 611-08-5 | C4H3N3O4 | 157.09 g/mol
Quantity:
25 g
100 g
Catalog number A12448.22
also known as A12448-22
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS611-08-5
IUPAC Name5-nitro-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC4H3N3O4
InChI KeyTUARVSWVPPVUGS-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CNC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder or needles
Assay from Supplier's CofA≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only