2-Chloro-6-nitrobenzonitrile, 98%
2-Chloro-6-nitrobenzonitrile, 98%
Thermo Scientific Chemicals

2-Chloro-6-nitrobenzonitrile, 98%

CAS: 6575-07-1 | C8H6ClN | 151.59 g/mol
Quantity:
5 g
25 g
Catalog number A12703.06
also known as A12703-06
Price (JPY)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS6575-07-1
IUPAC Name2-chloro-6-nitrobenzonitrile
Molecular FormulaC7H3ClN2O2
InChI KeyZHLCARBDIRRRHD-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC(Cl)=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to green to brown
FormCrystals or powder or crystalline powder
Melting Point (clear melt)118.0-124.0?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only