Thermo Scientific Chemicals

1,3-Dichloro-5,5-dimethylhydantoin, 98%, Thermo Scientific Chemicals

Catalog number: A12718.22
100 g, Each
Thermo Scientific Chemicals

1,3-Dichloro-5,5-dimethylhydantoin, 98%, Thermo Scientific Chemicals

Catalog number: A12718.22
100 g, Each
Quantity
Catalog number: A12718.22
also known as A12718-22
Price (USD)
Price: 25.30
Online price: 21.65
Your price:
Quantity
-

Chemical Identifiers

CAS
118-52-5
IUPAC Name
1,3-dichloro-5,5-dimethylimidazolidine-2,4-dione
Molecular Formula
C5H6Cl2N2O2
InChI Key
KEQGZUUPPQEDPF-UHFFFAOYSA-N
SMILES
CC1(C)N(Cl)C(=O)N(Cl)C1=O
Form
Powder
Identification (FTIR)
Conforms
Appearance (Color)
White
Assay (Iodometric Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
130-136?C

Description

Dantoin(R) DCDMH is an industrial bleaching agent based on dimethyl hydantoin. This product finds application wherever solid bleach is desired. It is used as a mild chlorinating agent in organic synthesis.It is also used as a sanitizer in swimming pools, laundries restaurants, cutting oils and pharmaceutical industry as a general cleaner. Other uses include it is been used as a preventer and catalyst in polymer industry, used to emboss or texturize resinous sheet preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dantoin(R) DCDMH is an industrial bleaching agent based on dimethyl hydantoin. This product finds application wherever solid bleach is desired. It is used as a mild chlorinating agent in organic synthesis.It is also used as a sanitizer in swimming pools, laundries restaurants, cutting oils and pharmaceutical industry as a general cleaner. Other uses include it is been used as a preventer and catalyst in polymer industry, used to emboss or texturize resinous sheet preparations.

Solubility
Soluble in water 0.21 g/100 mL (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with reducing agents, acids, strong bases, organic materials, powdered metals. Moisture, light, air sensitive. Stable, but a strong oxidizer.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text