3,5-Dibromobenzyl bromide, is an important raw material and intermediateintermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Dibromobenzyl bromide, is an important raw material and intermediateintermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in toluene
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to water, oxidizing agents, bases, alcohols, amines, steel.
RUO – Research Use Only
General References:
- CP Yang.; TM Lee. Syntheses and properties of 4-hydroxy-3, 5-dibromobenzyl phosphonates and their flame-retarding effects on ABS copolymer. Journal of Polymer Science Part A: Polymer. 1989 27, (7) , 2239-2251.
- R Epple.; M Azimioara.; R Russo.; B Bursulaya. 1, 3, 5-Trisubstituted aryls as highly selective PPARδ agonists. Bioorganic & Medicinal Chemistry Letters. 2006 16, (11) , 2969-2973.