CTS™ DPBS, calcium, magnesium
CTS™ DPBS, calcium, magnesium
CTS™ DPBS, calcium, magnesium
CTS™ DPBS, calcium, magnesium
Gibco™

CTS™ DPBS, calcium, magnesium

Gibco Cell Therapy Systems DPBS CTS is the classical Dulbecco's Phosphate Buffered Saline which contains calcium, magnesium but no phenolRead more
Have Questions?
Catalog NumberQuantity
A12858011000 mL
Catalog number A1285801
Price (USD)
70.65
Online exclusive
75.75
Save 5.10
Each
Quantity:
1000 mL
Price (USD)
70.65
Online exclusive
75.75
Save 5.10
Each

Gibco Cell Therapy Systems DPBS CTS is the classical Dulbecco's Phosphate Buffered Saline which contains calcium, magnesium but no phenol red.
Gibco DPBS CTS is manufactured in state-of-the-art cGMP, ISO certified facilities to help ensure the highest quality and consistency for reproducible results every day. Tests include osmolality, pH, stability, the absence of bacterial and fungal contamination and endotoxin tests.

Gibco CTS products are of high quality, and are supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate. The Gibco CTS product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.

This product will have the following intended use specific for cell therapy market - For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Chemical Name or MaterialDPBS
IncludesCalcium and Magnesium
Manufacturing QualityISO13485, MDSAP, FDA-registered, 21 CFR820
PackagingBottle
Recommended StorageStore at 2–8° C.

SterilitySterile-filtered
Concentration1X
For Use With (Application)Cell and Gene Therapy Research, Development, and Manufacturing
Physical FormLiquid
Quantity1000 mL
Solution TypeDulbecco's Phosphate Buffered Saline
Unit SizeEach

Frequently asked questions (FAQs)

What is CTS?

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.

Can I get a specific value for the conductivity for your DPBS products?

Sorry, we do not test the conductivity of our DPBS products and hence cannot provide a value or range.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are your DPBS products DNase/RNase-free? Can I use them for RNA work?

Our DPBS products are for cell culture use and have not been tested for DNase/RNase activity. We do not recommend using these products for RNA work.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (2)

Citations & References
Abstract
Plaque formation and isolation of pure lines with poliomyelitis viruses.
Authors:DULBECCO R, VOGT M
Journal:J Exp Med
PubMed ID:13130792
Plaques have been produced with the three types of poliomyelitis viruses on monolayer tissue cultures of monkey kidney and monkey testis. The number of plaques was proportional to the concentration of the virus. Each plaque originates, therefore, from a single virus particle, defined as the virus unit that is unseparable ... More
Chronic ethanol feeding increases the severity of Staphylococcus aureus skin infections by altering local host defenses.
Authors:Parlet CP, Kavanaugh JS, Horswill AR, Schlueter AJ
Journal:
PubMed ID:25605871
Alcoholics are at increased risk of Staphylococcus aureus skin infection and serious sequelae, such as bacteremia and death. Despite the association between alcoholism and severe S. aureus skin infection, the impact of EtOH on anti-S. aureus cutaneous immunity has not been investigated in a model of chronic EtOH exposure. To ... More