Methyl 4-nitrobenzoate, 99%
Methyl 4-nitrobenzoate, 99%
Methyl 4-nitrobenzoate, 99%
Thermo Scientific Chemicals

Methyl 4-nitrobenzoate, 99%

CAS: 619-50-1 | C8H7NO4 | 181.147 g/mol
Quantity:
100 g
500 g
Catalog number A12928.36
also known as A12928-36
Price (JPY)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS619-50-1
IUPAC Namemethyl 4-nitrobenzoate
Molecular FormulaC8H7NO4
InChI KeyYOJAHJGBFDPSDI-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystals or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)94-101?C
It is employed in the biochemical characterization of NfsA, the Escherichia coli major nitroreductase exhibiting a high amino acid sequence homology to Frp, a Vibrio harveyi flavin oxidoreductase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the biochemical characterization of NfsA, the Escherichia coli major nitroreductase exhibiting a high amino acid sequence homology to Frp, a Vibrio harveyi flavin oxidoreductase.

Solubility
Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. H. Wu, et al. Methyl 4-nitro­benzoate.Acta Cryst.,2009,E653096.
  2. Peter. J. Groenewegen. Novel degradative pathway of 4-nitrobenzoate in Comamonas acidovorans NBA-10.Journal of’ General Microbiolog.,1992,1381599-1605.