n-Heptylbenzene, 98%
n-Heptylbenzene, 98%
n-Heptylbenzene, 98%
Thermo Scientific Chemicals

n-Heptylbenzene, 98%

CAS: 1078-71-3 | C13H20 | 176.303 g/mol
Quantity:
25 g
100 g
500 g
Catalog number A12969.14
also known as A12969-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1078-71-3
IUPAC Nameheptylbenzene
Molecular FormulaC13H20
InChI KeyLBNXAWYDQUGHGX-UHFFFAOYSA-N
SMILESCCCCCCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.4840-1.4880 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only