Thermo Scientific Chemicals

L-(-)-Arabitol, 98%

Catalog number: A13103.22
100 g, Each
Thermo Scientific Chemicals

L-(-)-Arabitol, 98%

Catalog number: A13103.22
100 g, Each
Quantity
Catalog number: A13103.22
also known as A13103-22
Price (USD)
Price: 570.00
Online price: 484.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7643-75-6
IUPAC Name
pentane-1,2,3,4,5-pentol
Molecular Formula
C5H12O5
InChI Key
HEBKCHPVOIAQTA-UHFFFAOYNA-N
SMILES
OCC(O)C(O)C(O)CO
Appearance (Color)
White to pale cream
Assay (Silylated GC)
≥97.5%
Identification (FTIR)
Conforms
Optical Rotation
-9.0 ? 2.0? (C=5 in 4% in borax solution)
Form
Powder

Description

L-Arabitol, a rare sugar alcohol, is being studied as a food additive that reduces fat deposits in the intestines. L-Arabitol is used as an inducer of xylanase expression in Hypocrea jecorina (Trichoderma reesei). L-Arabitol is used to identify, differentiate and characterize L-arabitol dehydrogenase(s).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Arabitol, a rare sugar alcohol, is being studied as a food additive that reduces fat deposits in the intestines. L-Arabitol is used as an inducer of xylanase expression in Hypocrea jecorina (Trichoderma reesei). L-Arabitol is used to identify, differentiate and characterize L-arabitol dehydrogenase(s).

Solubility
Soluble in water (50 mg/ml).

Notes
It is hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text