2-Chloro-5-cyanopyridine, 97%
2-Chloro-5-cyanopyridine, 97%
2-Chloro-5-cyanopyridine, 97%
Thermo Scientific Chemicals

2-Chloro-5-cyanopyridine, 97%

CAS: 33252-28-7 | C6H3ClN2 | 138.554 g/mol
Quantity:
1 g
5 g
25 g
Catalog number A13291.03
also known as A13291-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS33252-28-7
IUPAC Name6-chloropyridine-3-carbonitrile
Molecular FormulaC6H3ClN2
InChI KeyORIQLMBUPMABDV-UHFFFAOYSA-N
SMILESClC1=CC=C(C=N1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)114.0-121.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only