4-Hydrazinobenzenesulfonic acid hydrate, 98%
4-Hydrazinobenzenesulfonic acid hydrate, 98%
4-Hydrazinobenzenesulfonic acid hydrate, 98%
Thermo Scientific Chemicals

4-Hydrazinobenzenesulfonic acid hydrate, 98%

CAS: 98-71-5 | C6H8N2O3S | 188.20 g/mol
Quantity:
5 g
25 g
100 g
Catalog number A13362.22
also known as A13362-22
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS98-71-5
IUPAC Name4-hydrazinylbenzene-1-sulfonic acid
Molecular FormulaC6H8N2O3S
InChI KeyIOMZCWUHFGMSEJ-UHFFFAOYSA-N
SMILESNNC1=CC=C(C=C1)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to pale brown
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (HPLC)≥97.5%
Identification (FTIR)Conforms
Loss on Drying≤19.31% (105?C/constant weight)
View more
4-Hydrazinobenzenesulfonic Acid is used as an intermediate to manufacture pharmaceuticals, dyes and other organic synthesis. They have active applications in organic synthesis for agrochemicals, pharmaceuticals, photographic, heat stabilizers, polymerization catalysts, flame-retardants, blowing agents for plastics, explosives, and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydrazinobenzenesulfonic Acid is used as an intermediate to manufacture pharmaceuticals, dyes and other organic synthesis. They have active applications in organic synthesis for agrochemicals, pharmaceuticals, photographic, heat stabilizers, polymerization catalysts, flame-retardants, blowing agents for plastics, explosives, and dyes.

Solubility
It is soluble in water.

Notes
Stable under normal conditions.Keep away from strong oxidizing agents. Store in closed vessels.Hygroscopic
RUO – Research Use Only
WW Bennett.; PR Teasdale.; JG Panther. New diffusive gradients in a thin film technique for measuring inorganic arsenic and selenium (IV) using a titanium dioxide based adsorbent. Anal. Chem. 2010, 82,(7), 7401-7407.