1,8-Naphthalic anhydride, 97%
1,8-Naphthalic anhydride, 97%
1,8-Naphthalic anhydride, 97%
Thermo Scientific Chemicals

1,8-Naphthalic anhydride, 97%

CAS: 81-84-5 | C12H6O3 | 198.18 g/mol
Quantity:
50 g
250 g
1000 g
Catalog number A13519.18
also known as A13519-18
Price (JPY)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS81-84-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to pale brown
CommentPurchased in U.K. and U.S.
Identification (FTIR)Conforms (U.K. specification)
Assay (HPLC)≥96.0%
FormPowder or crystalline powder
View more
1,8-Naphthalic anhydride is used as a seed protectant and to prepare dyestuffs and optical brighteners. It is used in the preparation of napthalimide analogues, which finds application as anticancer agents. Further, it is used as fluorescent brightening agent for polymeric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,8-Naphthalic anhydride is used as a seed protectant and to prepare dyestuffs and optical brighteners. It is used in the preparation of napthalimide analogues, which finds application as anticancer agents. Further, it is used as fluorescent brightening agent for polymeric materials.

Solubility
Soluble in ether and alcohol. Slightly soluble in acetic acid. Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only

General References:

  1. Wang, F.; Sun, J.; Wang, J.; Bai, S.; Wu, X. Influence of Various Solvents on the Luminescent Performance of 1,8-Naphthalic Anhydride Modified by Eu3+ Ions. J. Nanosci. Nanotechnol. 2015, 15 (6), 4347-4352.
  2. Gerbich, T.; Schmitt, H. C.; Fischer, I.; Petersen, J.; Albert, J.; Mitrić, R. Time-Resolved Study of 1,8-Naphthalic Anhydride and 1,4,5,8-Naphthalene-tetracarboxylic Dianhydride. J. Phys. Chem. A 2015, 119 (23), 6006-6016.