2-Chloro-N,N-diethylacetamide, 97%
2-Chloro-N,N-diethylacetamide, 97%
2-Chloro-N,N-diethylacetamide, 97%
Thermo Scientific Chemicals

2-Chloro-N,N-diethylacetamide, 97%

CAS: 2315-36-8 | C6H12ClNO | 149.618 g/mol
Quantity:
5 g
25 g
100 g
Catalog number A13691.22
also known as A13691-22
Price (JPY)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS2315-36-8
IUPAC Name2-chloro-N,N-diethylacetamide
Molecular FormulaC6H12ClNO
InChI KeyCQQUWTMMFMJEFE-UHFFFAOYSA-N
SMILESCCN(CC)C(=O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow or pale pink
FormLiquid
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.4680-1.4730 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only