4-Nitro-3-(trifluoromethyl)aniline, 98%
4-Nitro-3-(trifluoromethyl)aniline, 98%
4-Nitro-3-(trifluoromethyl)aniline, 98%
Thermo Scientific Chemicals

4-Nitro-3-(trifluoromethyl)aniline, 98%

CAS: 393-11-3 | C7H5F3N2O2 | 206.12 g/mol
Quantity:
1 g
5 g
Catalog number A13798.03
also known as A13798-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS393-11-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)122.0-128.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only