3,4,5-Triethoxybenzoic acid, 98+%
3,4,5-Triethoxybenzoic acid, 98+%
3,4,5-Triethoxybenzoic acid, 98+%
Thermo Scientific Chemicals

3,4,5-Triethoxybenzoic acid, 98+%

CAS: 6970-19-0 | C13H18O5 | 254.282 g/mol
Quantity:
25 g
100 g
500 g
Catalog number A14157.14
also known as A14157-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS6970-19-0
IUPAC Name3,4,5-triethoxybenzoic acid
Molecular FormulaC13H18O5
InChI KeyYCENSLDYFDZUMO-UHFFFAOYSA-N
SMILESCCOC1=CC(=CC(OCC)=C1OCC)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)111.0-117.0?C
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Appearance (Color)White to pale cream
Assay (HPLC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only