2,2,2-Trifluoroacetamide, 97%
2,2,2-Trifluoroacetamide, 97%
2,2,2-Trifluoroacetamide, 97%
Thermo Scientific Chemicals

2,2,2-Trifluoroacetamide, 97%

CAS: 354-38-1 | C2H2F3NO | 113.04 g/mol
Quantity:
25 g
100 g
500 g
Catalog number A14451.14
also known as A14451-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS354-38-1
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)68.0-77.0?C
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
2,2,2-Trifluoroacetamide used in a convenient alternative to the Gabriel synthesis of primary amines from halides by N-alkylation followed by cleavage of the readily-hydrolyzed trifluoroacetyl group and for improved N-alkylation under phase-transfer conditions allowing successive alkylation with different alkyl groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trifluoroacetamide used in a convenient alternative to the Gabriel synthesis of primary amines from halides by N-alkylation followed by cleavage of the readily-hydrolyzed trifluoroacetyl group and for improved N-alkylation under phase-transfer conditions allowing successive alkylation with different alkyl groups.

Solubility
Soluble in water (460 g/L at 20°C), alcohol, ether, chloroform (partially), and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
RUO – Research Use Only

General References:

  1. Chuan-Zhou Tao.; Juan Li.; Yao Fu.; Lei Liu.; Qing-Xiang Guo. Copper-catalyzed synthesis of primary arylamines from aryl halides and 2,2,2-trifluoroacetamide. Tetrahedron Letters. 2008, 49 (1), 70-75.
  2. Snefrid Gundersen.; Svein Samdal.; Ragnhild Seip.; Dmitry J. Shorokhov.; Tor G. Strand. Applequist. The molecular structure, conformation, potential to internal rotation and force field of 2,2,2-trifluoroacetamide as studied by gas electron diffraction and quantum chemical calculations. Journal of Molecular Structure. 1998, 445 (1-3), 229-242.
  3. Has been used in a convenient alternative to the Gabriel synthesis of primary amines from halides by N-alkylation followed by cleavage of the readily-hydrolyzed trifluoroacetyl group: Synthesis, 941 (1984). For improved N-alkylation under phase-transfer conditions allowing successive alkylation with different alkyl groups, see: Synth. Commun., 18, 791 (1988). The method has been further extended by the use of ɑ-bromo esters to provide a high yielding route to ɑ-amino acids: J. Org. Chem., 56, 420 (1991):