N,N-Dimethylmethanesulfonamide, 98+%
N,N-Dimethylmethanesulfonamide, 98+%
N,N-Dimethylmethanesulfonamide, 98+%
Thermo Scientific Chemicals

N,N-Dimethylmethanesulfonamide, 98+%

CAS: 918-05-8 | C3H9NO2S | 123.17 g/mol
Quantity:
5 g
25 g
Catalog number A14454.06
also known as A14454-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS918-05-8
IUPAC NameN,N-dimethylmethanesulfonamide
Molecular FormulaC3H9NO2S
InChI KeyWCFDSGHAIGTEKL-UHFFFAOYSA-N
SMILESCN(C)S(C)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale pink
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥98.0%
Melting Point (clear melt)42.0-51.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only