3-Amino-1H-pyrazole, 97+%
3-Amino-1H-pyrazole, 97+%
3-Amino-1H-pyrazole, 97+%
Thermo Scientific Chemicals

3-Amino-1H-pyrazole, 97+%

CAS: 1820-80-0 | C3H5N3 | 83.094 g/mol
Quantity:
1 g
5 g
25 g
Catalog number A14458.03
also known as A14458-03
Price (JPY)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1820-80-0
IUPAC Name1H-pyrazol-5-amine
Molecular FormulaC3H5N3
InChI KeyJVVRJMXHNUAPHW-UHFFFAOYSA-N
SMILESNC1=CC=NN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to brown
FormLiquid or fused solid
Assay (GC)≥97.0%
Melting Point (clear melt)31-39°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only