3-Octen-2-one, 97%
3-Octen-2-one, 97%
3-Octen-2-one, 97%
Thermo Scientific Chemicals

3-Octen-2-one, 97%

CAS: 1669-44-9 | C8H14O | 126.199 g/mol
Quantity:
10 g
50 g
250 g
Catalog number A14652.09
also known as A14652-09
Price (JPY)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS1669-44-9
IUPAC Name(3E)-oct-3-en-2-one
Molecular FormulaC8H14O
InChI KeyZCFOBLITZWHNNC-VOTSOKGWSA-N
SMILESCCCCC=CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Colorless to pale yellow
Refractive Index1.4455-1.4505 @ 20?C
FormLiquid
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only