Thermo Scientific Chemicals

n-Tetradecane, 99%, Thermo Scientific Chemicals

Catalog number: A14672.7W
1500 g, Each
Thermo Scientific Chemicals

n-Tetradecane, 99%, Thermo Scientific Chemicals

Catalog number: A14672.7W
1500 g, Each
Quantity
Catalog number: A14672.7W
also known as A14672-7W
Price (USD)
Price: 705.00
Online price: 599.65
Your price:
Quantity
-

Chemical Identifiers

CAS
629-59-4
IUPAC Name
tetradecane
Molecular Formula
C14H30
InChI Key
BGHCVCJVXZWKCC-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCC
Appearance (Color)
Clear, colorless
Assay (GC)
≥98.5%
Form
Liquid

Description

n-Tetradecane is used to study the thermodiffusion behavior of the microemulsion droplets of water, n-alkanes and pentaethylene glycol monododecyl ether. It is used as reference material in gas chromatographic analysis, as a solvent and finds applications in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Tetradecane is used to study the thermodiffusion behavior of the microemulsion droplets of water, n-alkanes and pentaethylene glycol monododecyl ether. It is used as reference material in gas chromatographic analysis, as a solvent and finds applications in organic synthesis.

Solubility
Soluble in ether, acetone and alcohol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text