Phthalonitrile, 98%
Phthalonitrile, 98%
Phthalonitrile, 98%
Thermo Scientific Chemicals

Phthalonitrile, 98%

CAS: 91-15-6 | C8H4N2 | 128.13 g/mol
Quantity:
100 g
250 g
1000 g
Catalog number A15071.30
also known as A15071-30
Price (JPY)
-
Request A Quote
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS91-15-6
IUPAC Namebenzene-1,2-dicarbonitrile
Molecular FormulaC8H4N2
InChI KeyXQZYPMVTSDWCCE-UHFFFAOYSA-N
SMILESN#CC1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White or pale cream to pale brown
FormPowder or flakes
Melting Point (clear melt)136-142?C
Assay (GC)≥97.5%
Phthalonitrile is used as an intermediate for chemical synthesis in the production of building blocks for colorants and coatings, life science and agricultural chemicals. And also used as a precursor to phthalocyanine and other pigments, fluorescent brighteners, and photographic sensitizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalonitrile is used as an intermediate for chemical synthesis in the production of building blocks for colorants and coatings, life science and agricultural chemicals. And also used as a precursor to phthalocyanine and other pigments, fluorescent brighteners, and photographic sensitizers.

Solubility
Soluble in alcohol and ether.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Sergiu M. Gorun.; Barbara A. Bench.; Gene Carpenter.; Matthew W. Beggs.; Joel T. Mague.; Harry E. Ensley. Synthesis and structural characterization of non-planar perfluoro phthalonitriles. Journal of Fluorine Chemistry. 1998, 91, (1), 37-40.
  2. Rodolfo Nesi.; Stefania Turchi.; Donatella Giomi.; Camilla Corsi. Phthalonitriles by hetero Diels-Alder reactions of 4,5-dicyanopyridazine with enamines: isolation and characterization of unprecedented intermediates. Tetrahedron. 1998, 54, (36), 10851-10856.