3-Cyclohexene-1-carboxylic acid, 98%
3-Cyclohexene-1-carboxylic acid, 98%
3-Cyclohexene-1-carboxylic acid, 98%
Thermo Scientific Chemicals

3-Cyclohexene-1-carboxylic acid, 98%

CAS: 4771-80-6 | C7H10O2 | 126.155 g/mol
Quantity:
5 g
25 g
Catalog number A15229.14
also known as A15229-14
Price (JPY)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS4771-80-6
IUPAC Namecyclohex-3-ene-1-carboxylic acid
Molecular FormulaC7H10O2
InChI KeyVUSWCWPCANWBFG-UHFFFAOYNA-N
SMILESOC(=O)C1CCC=CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Refractive Index1.4785-1.4825 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only