3-Chloro-4-methylaniline, 98%
3-Chloro-4-methylaniline, 98%
3-Chloro-4-methylaniline, 98%
3-Chloro-4-methylaniline, 98%
Thermo Scientific Chemicals

3-Chloro-4-methylaniline, 98%

CAS: 95-74-9 | C7H8ClN | 141.598 g/mol
Quantity:
250 g
1000 g
Catalog number A15439.0B
also known as A15439-0B
Price (JPY)
-
Request A Quote
Quantity:
1000 g
Request bulk or custom format
Chemical Identifiers
CAS95-74-9
IUPAC Name3-chloro-4-methylaniline
Molecular FormulaC7H8ClN
InChI KeyRQKFYFNZSHWXAW-UHFFFAOYSA-N
SMILESCC1=CC=C(N)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear red to brown
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.5820-1.5860 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only