Thermo Scientific Chemicals

3-Nitrophthalic anhydride, 97%, Thermo Scientific Chemicals

Catalog number: A15857.14
25 g, Each
Thermo Scientific Chemicals

3-Nitrophthalic anhydride, 97%, Thermo Scientific Chemicals

Catalog number: A15857.14
25 g, Each
Quantity
Catalog number: A15857.14
also known as A15857-14
Price (USD)
Price: 136.00
Online price: 115.65
Your price:
Quantity
-

Chemical Identifiers

CAS
641-70-3
IUPAC Name
4-nitro-1,3-dihydro-2-benzofuran-1,3-dione
Molecular Formula
C8H3NO5
InChI Key
ROFZMKDROVBLNY-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=CC2=C1C(=O)OC2=O
Melting Point (clear melt)
160-166°C
Appearance (Color)
White to yellow to pale brown or cream
Form
Crystalline powder
Assay (Aqueous acid-base Titration)
≥96.0%

Description

4-Nitrophthalic Anhydride is an intermediate for the synthesis of the benzimidazole PARP inhibitor I, ABT-472. It reacts with aminoquinazolinones to yield phthalimidoquinazolinones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophthalic Anhydride is an intermediate for the synthesis of the benzimidazole PARP inhibitor I, ABT-472. It reacts with aminoquinazolinones to yield phthalimidoquinazolinones.

Solubility
May decompose in water.

Notes
Air sensitive. Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text