Thermo Scientific Chemicals

Mercury(I) nitrate dihydrate, 97%, Thermo Scientific Chemicals

Catalog number: A16268.30
250 g, Each
Thermo Scientific Chemicals

Mercury(I) nitrate dihydrate, 97%, Thermo Scientific Chemicals

Catalog number: A16268.30
250 g, Each
Quantity
Catalog number: A16268.30
also known as A16268-30
Price (USD)

Chemical Identifiers

CAS
7782-86-7
IUPAC Name
mercury(1+) hydrate nitrate
Molecular Formula
H2HgNO4
InChI Key
UPCSQZXLTBTBCU-UHFFFAOYSA-N
SMILES
O.[Hg+].[O-][N+]([O-])=O
Comment
May contain small amounts of dark particulate matter as part of manufacturing process
Appearance (Color)
White to yellow to yellow-red
Form
Crystalline or lumpy powder
Assay (unspecified)
≥96.0%

Description

Mercury(I) nitrate dihydrate is used in the preparation of other mercury(I) compounds. It acts as a reducing agent. It is involved in the preparation of mercury(II) nitrate by treating with dilute nitric acid as well as to get mercury on exposure to light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mercury(I) nitrate dihydrate is used in the preparation of other mercury(I) compounds. It acts as a reducing agent. It is involved in the preparation of mercury(II) nitrate by treating with dilute nitric acid as well as to get mercury on exposure to light.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with alcohols, sulfur and strong reducing agents.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text