3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, 97%
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, 97%
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, 97%
Thermo Scientific Chemicals

3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, 97%

CAS: 149022-15-1 | C8H10ClN3S | 215.70 g/mol
Quantity:
5 g
25 g
100 g
Catalog number A16317.06
also known as A16317-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS149022-15-1
IUPAC Name2-hydrazinylidene-3-methyl-2,3-dihydro-1,3-benzothiazole hydrochloride
Molecular FormulaC8H10ClN3S
InChI KeyOEZPVSPULCMUQB-UHFFFAOYSA-N
SMILESCl.CN1C(SC2=CC=CC=C12)=NN
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Identification (FTIR)Conforms
FormPowder
Appearance (Color)White to cream or pale pink
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
View more
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate is used in the synthesis of benzothiazolium azo dyes. It is a chromogenic reagent used for the determination of cholesterol, benzodiazepines and enzyme activity. As an electrophilic coupling reagent, it is used in the spectrophotometric determination of chloramine-B (CAB) and other residual chlorine content in various environmental water samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-benzothiazolinone hydrazone hydrochloride hydrate is used in the synthesis of benzothiazolium azo dyes. It is a chromogenic reagent used for the determination of cholesterol, benzodiazepines and enzyme activity. As an electrophilic coupling reagent, it is used in the spectrophotometric determination of chloramine-B (CAB) and other residual chlorine content in various environmental water samples.

Solubility
Soluble in water, dimethyl sulfoxide and methanol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Sensitive color reagent for the detection and estimation of water-soluble aldehydes: Anal. Chem., 33, 93, 722, 1574 (1961).
  2. Shi, J. Y; Liu, N.; Gu, R. X.; Zhu, L. Q.; Zhang, C.; Wang, Q. G.; Lei, Z. H.; Liu, Y. Y.; Ren, J. Y. Signals induced by exogenous nitric oxide and their role in controlling brown rot disease caused by Monilinia fructicola in postharvest peach fruit. J. Gen. Plant Pathol. 2015, 81 (1), 68-76.
  3. El-Sayed, A. S. A.; Abdel-Azeim, S.; Ibrahim, H. M.; Yassin, M. A.; Abdel-Ghany, S. E.; Esener, S.; Ali, G. S. Biochemical stability and molecular dynamic characterization of Aspergillus fumigatus cystathionine γ-lyase in response to various reaction effectors. Enzyme Microb. Technol. 2015, 81, 31-46.