2',3'-Difluoroacetophenone, 97%
2',3'-Difluoroacetophenone, 97%
2',3'-Difluoroacetophenone, 97%
Thermo Scientific Chemicals

2',3'-Difluoroacetophenone, 97%

CAS: 18355-80-1 | C8H6F2O | 156.13 g/mol
Quantity:
5 g
25 g
Catalog number A16786.06
also known as A16786-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS18355-80-1
IUPAC Name1-(2,3-difluorophenyl)ethan-1-one
Molecular FormulaC8H6F2O
InChI KeyPQUXFUBNSYCQAL-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.4875-1.4925 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only