1,2-Cyclohexanedione dioxime, 97%
1,2-Cyclohexanedione dioxime, 97%
1,2-Cyclohexanedione dioxime, 97%
1,2-Cyclohexanedione dioxime, 97%
Thermo Scientific Chemicals

1,2-Cyclohexanedione dioxime, 97%

CAS: 492-99-9 | C6H10N2O2 | 142.16 g/mol
Quantity:
5 g
25 g
100 g
Catalog number A16788.14
also known as A16788-14
Price (JPY)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS492-99-9
IUPAC NameN-[(1E,2E)-2-(hydroxyimino)cyclohexylidene]hydroxylamine
Molecular FormulaC6H10N2O2
InChI KeyCUNNCKOPAWXYDX-KQQUZDAGSA-N
SMILESO\N=C1/CCCC/C/1=N\O
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental AnalysisC: 50.69 ? 2%
Elemental AnalysisN: 19.71 ? 0.7%
Formpowder
Appearance (Color)White to pale brown
1,2-Cyclohexanedione dioxime acts as a chelating ligand forming complexes with many transition metal ions like Co, Fe and Ni. It is used in photometric detection of metal ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Cyclohexanedione dioxime acts as a chelating ligand forming complexes with many transition metal ions like Co, Fe and Ni. It is used in photometric detection of metal ions.

Solubility
Soluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. W. C. Johnson and M. Simmons. 1:2-Cyclohexanedione-dioxime (‘Nioxime’) as a reagent for nickel. Analyst.1946, 71 (1), 554-556.
  2. Chelating ligand forming complexes with many transition metal ions: J. Inorg. Nucl. Chem., 36, 1601 (1974); J. Am. Chem. Soc., 97, 932 (1975).