3,5-Difluorobenzaldehyde, 97%
3,5-Difluorobenzaldehyde, 97%
3,5-Difluorobenzaldehyde, 97%
Thermo Scientific Chemicals

3,5-Difluorobenzaldehyde, 97%

CAS: 32085-88-4 | C7H4F2O | 142.105 g/mol
Quantity:
1 g
5 g
25 g
Catalog number A16917.06
also known as A16917-06
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS32085-88-4
IUPAC Name3,5-difluorobenzaldehyde
Molecular FormulaC7H4F2O
InChI KeyASOFZHSTJHGQDT-UHFFFAOYSA-N
SMILESFC1=CC(C=O)=CC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Free acid (titration)≤3.5%
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Refractive Index1.4905-1.4955 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only